missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-11, Animal-Free Recombinant Protein, PeproTech®

Product Code. 17846538
Click to view available options
Quantity:
2 μg
250 μg
50 μg
Unit Size:
250µg
2µg
50µg
This item is not returnable. View return policy

Product Code. 17846538

Brand: Gibco™ AF200112UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human IL-11 is a 19.3 kDa protein consisting of 179 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Interleukins (ILs) are a large group of cytokines that are produced mainly by leukocytes, although some are produced by certain phagocytes and auxiliary cells. ILs have a variety of functions, but most function to direct other immune cells to divide and differentiate. Each IL acts on a specific, limited group of cells through a receptor specific for that IL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P20809
For Use With (Application) Functional Assay
Format Lyophilized
Formulation protein with no preservative
Gene ID (Entrez) 3589
Molecular Weight (g/mol) 19.3 kDa
Name Human IL-11, Animal-Free
Purification Method HPLC, SDS-PAGE
Quantity 2 μg
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/ μg
Gene Alias adipogenesis inhibitory factor; AGIF; il 11; IL10RA; Il11; IL-11; IL11 inhibitor; IL-11 ra; ILN; Interleukin; interleukin 11; Interleukin11; interleukin-11; MGC2146; Oprelvekin
Common Name IL-11
Gene Symbol IL11
Biological Activity The ED50 was determined by the dose-dependent stimulation of the proliferation of murine T11 cells is ≤ 2.0 ng/ml, corresponding to a specific activity of ≥ 5 x 10^5 units/mg
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MPGPPPGPPR VSPDPRAELD STVLLTRSLL ADTRQLAAQL RDKFPADGDH NLDSLPTLAM SAGALGALQL PGVLTRLRAD LLSYLRHVQW LRRAGGSSLK TLEPELGTLQ ARLDRLLRRL QLLMSRLALP QPPPDPPAPP LAPPSSAWGG IRAAHAILGG LHLTLDWAVR GLLLLKTRL
Content And Storage -20°C
Activity The ED50 was determined by the dose-dependent stimulation of the proliferation of murine T11 cells is ≤ 2.0 ng/ml, corresponding to a specific activity of ≥ 5 x 10^5 units/mg
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.