missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IL-27 Recombinant Protein, PeproTech®
HEK293 cells Recombinant Protein
Brand: Gibco™ 200-38-1MG
Additional Details : Weight : 0.16500kg
Description
IL-27 (Interleukin 27) is a cytokine belonging to the IL-12 family, and is a heterodimeric cytokine composed of EBI3 and p28 subunits. IL-27 is produced by activated dendritic cells and macrophages in response to Toll-like receptor ligands and pro-inflammatory cytokines. IL-27 promotes CD4+ T cell differentiation to the Th1 lineage by inducing expression of the transcription factor T-bet and up regulating IL-12R beta2, which in turn suppresses Th2 and Th17 differentiation and proliferation. IL-27 is a recently discovered member of the IL-6/IL-12 family of proinflammatory and immunoregulatory cytokines. IL-27 acts by binding to its receptor WSX-1 and gp130 which results in the activation of a Jak/STAT signaling cascade, suggesting IL-27 involvement in the regulation of immune processes. It has been suggested that IL-27 can also be used as a therapeutic agent against cancer as it can also induce tumor-specific anti-tumor activity mediated through CD8+ T-cells, IFN-gamma, and T-bet. In humans, the gene is located on the p arm of chromosome 16.Specifications
Q14213, Q8NEV9 | |
protein with no preservative | |
47.8 | |
1 mg | |
-20°C | |
<1 EU/μg | |
Ebi3, IL27 | |
Unconjugated | |
p28 subunit: FPRPPGRPQL SLQELRREFT VSLHLARKLL SEVRGQAHRF AESHLPGVNL YLLPLGEQLP DVSLTFQAWR RLSDPERLCF ISTTLQPFHA LLGGLGTQGR WTNMERMQLW AMRLDLRDLQ RHLRFQVLAA GFNLPEEEEE EEEEEEEERK GLLPGALGSA LQGPAQVSWP QLLSTYRLLH SLELVLSRAV RELLLLSKAG HSVWPLGFPT LSPQP EBI3 subunit: RKGPPAALTL PRVQCRASRY PIAVDCSWTL PPAPNSTSPV SFIATYRLGM AARGHSWPCL QQTPTSTSCT ITDVQLFSMA PYVLNVTAVH PWGSSSSFVP FITEHIIKPD PPEGVRLSPL AERQLQVQWE PPGSWPFPEI FSLKYWIRYK RQGAARFHRV GPIEATSFIL RAVRPRARYY VQVAAQDLTD YGELSDWSLP ATATMSLGK | |
≥ 90% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
10148, 246778 | |
Human IL-27 | |
HEK293 cells | |
RUO | |
cytokine receptor; EBI3; EBI-3; EBV-induced gene 3 protein; Epstein-Barr virus induced 3; Epstein-Barr virus induced gene 3; Epstein-Barr virus induced gene 3 (similar to Interleukin-27 beta chain precursor); epstein-Barr virus-induced gene 3 protein; Epstein-Barr virus-induced gene 3 protein homolog; Il27; IL-27; IL27 p28; IL-27 p28 subunit; IL27 subunit; IL-27 subunit alpha; IL-27 subunit beta; Il27a; IL-27 A; IL-27-A; IL27-A; Il27b; IL-27 B; IL27p28; IL-27p28; Il30; IL35 subunit; ILN; Interleukin; interleukin 27; interleukin 30; interleukin-27 subunit alpha; Interleukin-27 subunit beta; interleukin-30; MGC71873; M-IL-27 (p28); p28; RGD1561420 | |
Human | |
Recombinant | |
Lyophilized |