missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ Human IL-6R ELISA Kit

Product Code. 16861526 Shop All Proteintech Products
Change view
Click to view available options
Target Species:
Human
Unit Size:
96 tests
Product Code. Target Species unitSize
16861526 Human 96 tests
1 options
This item is not returnable. View return policy

Product Code. 16861526

Brand: Proteintech™ KE0011896T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Solid phase sandwich ELISA used to detect and quantify protein levels of endogenous IL6R

  • Recognizes human IL6R
  • IL-6 (Interleukin-6) is a pleiotropic cytokine produced by a variety of cells during infection, trauma and immunological challenge
    • Acts via receptor complex consisting of two distinct membrane-bound glycoproteins, 80 kDa IL-6 binding subunit (IL-6R) and 130 kDa signal-transducing element (gp130)
    • After binding IL-6 to membrane-bound IL-6R, complex of IL-6 adn IL-6R associates with gp130 activating the receptor
      • Expression of gp130 found in almost all organs while expression of IL-6R predominantly confied to hapatocytesand leukocyte subpopulations
      • Soluble form of IL-6R found in human serum and urine

Process:

  • Antibody specific for IL6R pre-coated onto microwells
  • IL6R protein in samples captured by the coated antibody after incubation
  • After extensive washing, another antibody of biotinylated specific for IL6R is added to detect captured IL6R protein
  • For signal development, Streptavidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent
  • Solution containing sulfuric acid used to stop color development and color intensity
    • Proportional to quantity of bound protein, measurable at 450 nm with correction wavelength set at 630 nm

Specifications

Accession Number BC132684
Assay Range 7.8 to 500 pg/mL
Assay Sensitivity 0.1 pg/mL
Certifications/Compliance ISO 13485
Detection Method Sandwich ELISA, Double Antibody
Immunoassay Kit Format Coated ELISA Kit
Sample Type Plasma, Serum
Substrate DAB
Target Species Human
Gene ID (Entrez) 3570
Host Species Human
Immunoassay Kit Grade Solid
Interassay CV <10%
Intraassay CV <10%
Shipping Condition Blue ice
Storage Requirements -20°C to +4°C
Total Assay Time 4.5 hr.
UniProt ID P08887
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 96TEST Human IL-6R ELISA Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Sulphuric Acid, Tetramethylbenzidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.