Learn More
Invitrogen™ Human IRS1 (aa 631-646) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP185
Additional Details : CAS : 69-74-9 Weight : 0.01000kg
Description
Human IRS-1 (residues 631-646) Synthetic Peptide, PEP-185, from Invitrogen.
PEP-185 is a 16 amino acid synthetic peptide corresponding to amino acid residues 631-646 from human IRS-1. The sequence of this peptide is (amino to carboxy terminus): D Y M P M (pS) P K S V S A P Q Q I (NH2).This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product, catalog # PA1-1055. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Specifications
3667 | |
purified | |
Human IRS1 (aa 631-646) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
P35568 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
IRS1 | |
No preservative | |
50 μg | |
D Y M P M (pS) P K S V S A P Q Q I(NH2) |