Learn More
Invitrogen™ Human ITIH1 Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP301
Additional Details : Weight : 0.99000kg
Description
ITI-HC1 Synthetic Peptide, PEP-301, from Invitrogen™.
The inter-alpha-trypsin inhibitors (ITI) are a family of structurally related plasma serine protease inhibitors involved in extracellular matrix stabilization and in prevention of tumor metastasis. The ITI family contains multiple proteins made up of a light chain (see MIM 176870) and a variable number of heavy chains (Salier et al., 1987 [PubMed 2446322]; Himmelfarb et al., 2004 [PubMed 14744536]). This peptide corresponds to amino acids Q(383) VQESLPELSN (393) of human ITI-HC1 and can be used as a blocking peptide with polyclonal antibody PA1-1755.Specifications
3697 | |
purified | |
Human ITIH1 | |
-20°C | |
Lyophilized | |
Blocking Assay, Control | |
ITIH1 | |
No preservative | |
50 μg |
Human | |
P19827 | |
1 mg/mL | |
Synthetic Peptide | |
water with no preservative | |
CYCSP39, cytochrome c, somatic pseudogene 39, HCP39 | |
Yes | |
Peptide | |
RUO - research use only |