missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Human Kidney Whole Tissue Lysate (Adult Whole Primary Tumor and Normal Paired)
Shop All Bio Techne Products
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
Description
The Human Kidney Whole Tissue Lysate (Adult Whole Primary Tumor and Normal Paired) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.
Specifications
Specifications
| For Use With (Application) | Western Blot |
| Tissue | Digestive/Waste |
| Quantity | 1 Set |
| Storage Buffer | HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors |
| Test Specificity | Matched Pair - Total Protein - Human Primary Tumor and Normal Tissue (PP): Kidney |
| Cellular Fraction | Whole |
| Tissue Condition | Primary Tumor and Normal Paired |
| Cross Reactivity | Human |
| Host Species | Human |
| Concentration | 5 mg/mL |
| Show More |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction