missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LAMA3 Partial ORF (NP_000218, 29 a.a. - 138 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00003909-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16157291

  • 528.00€ / 25µg
Estimated Shipment: 09-11-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Laminins are basement membrane components thought to mediate the attachment, migration and organization of cells into tissues during embryonic development by interacting with other extracellular matrix components. The protein encoded by this gene is the alpha-3 subunit of laminin 5, which is a complex glycoprotein composed of three subunits (alpha, beta, and gamma). Laminin 5 is thought to be involved in cell adhesion, signal transduction and differentiation of keratinocytes. Mutations in this gene have been identified as the cause of Herlitz type junctional epidermolysis bullosa. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene. [provided by RefSeq]

Sequence: SSQQQRVPFLQPPGQSQLQASYVEFRPSQGCSPGYYRDHKGLYTGRCVPCNCNGHSNQCQDGSGICVNCQHNTAGEHCERCQEGYYGNAVHGSCRACPCPHTNSFATGCV
Specifications

Specifications

NP_000218
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SSQQQRVPFLQPPGQSQLQASYVEFRPSQGCSPGYYRDHKGLYTGRCVPCNCNGHSNQCQDGSGICVNCQHNTAGEHCERCQEGYYGNAVHGSCRACPCPHTNSFATGCV
RUO
LAMA3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
3909
LAMA3 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BM600/E170/LAMNA/LOCS/lama3a
LAMA3
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions