missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LGALS2 Full-length ORF (AAH59782.1, 1 a.a. - 132 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00003957-P02.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16157371

  • 349.00€ / 10µg
Estimated Shipment: 09-11-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is a soluble beta-galactoside binding lectin. The encoded protein is found as a homodimer and can bind to lymphotoxin-alpha. A single nucleotide polymorphism in an intron of this gene can alter the transcriptional level of the protein, with a resultant increased risk of myocardial infarction. [provided by RefSeq]

Sequence: MTGELEVKNMDMKPGSTLKITGSIADGTDGFVINLGQGTDKLNLHFNPRFSESTIVCNSLDGSNWGQEQREDHLCFSPGSEVKFTVTFESDKFKVKLPDGHELTFPNRLGHSHLSYLSVRGGFNMSSFKLKE
Specifications

Specifications

AAH59782.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
40.92kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HL14/MGC75071
LGALS2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
3957
LGALS2 (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue
MTGELEVKNMDMKPGSTLKITGSIADGTDGFVINLGQGTDKLNLHFNPRFSESTIVCNSLDGSNWGQEQREDHLCFSPGSEVKFTVTFESDKFKVKLPDGHELTFPNRLGHSHLSYLSVRGGFNMSSFKLKE
RUO
LGALS2
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions