missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LIG1 Partial ORF (NP_000225, 810 a.a. - 919 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00003978-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16157391

  • 528.00€ / 25µg
Estimated Shipment: 10-11-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

LIG1 encodes DNA ligase I, with functions in DNA replication and the base excision repair process. Mutations in LIG1 that lead to DNA ligase I deficiency result in immunodeficiency and increased sensitivity to DNA-damaging agents. [provided by RefSeq]

Sequence: QSLKALVLPSPRPYVRIDGAVIPDHWLDPSAVWEVKCADLSLSPIYPAARGLVDSDKGISLRFPRFIRVREDKQPEQATTSAQVACLYRKQSQIQNQQGEDSGSDPEDTY
Specifications

Specifications

NP_000225
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
QSLKALVLPSPRPYVRIDGAVIPDHWLDPSAVWEVKCADLSLSPIYPAARGLVDSDKGISLRFPRFIRVREDKQPEQATTSAQVACLYRKQSQIQNQQGEDSGSDPEDTY
RUO
LIG1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
3978
LIG1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC117397/MGC130025
LIG1
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions