missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LOH12CR1 Full-length ORF (NP_477517.1, 1 a.a. - 196 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00118426-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16157423

  • 528.00€ / 25µg
Estimated Shipment: 09-11-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MGSEQSSEAESRPNDLNSSVTPSPAKHRAKMDDIVVVAQGSQASRNVSNDPDVIKLQEIPTFQPLLKGLLSGQTSPTNAKLEKLDSQQVLQLCLRYQDHLHQCAEAVAFDQNALVKRIKEMDLSVETLFSFMQERQKRYAKYAEQIQKVNEMSAILRRIQMGIDQTVPLLDRLNSMLPEGERLEPFSMKPDRELRL
Specifications

Specifications

NP_477517.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
48.6kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
LOH1CR12
LOH12CR1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
118426
LOH12CR1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGSEQSSEAESRPNDLNSSVTPSPAKHRAKMDDIVVVAQGSQASRNVSNDPDVIKLQEIPTFQPLLKGLLSGQTSPTNAKLEKLDSQQVLQLCLRYQDHLHQCAEAVAFDQNALVKRIKEMDLSVETLFSFMQERQKRYAKYAEQIQKVNEMSAILRRIQMGIDQTVPLLDRLNSMLPEGERLEPFSMKPDRELRL
RUO
LOH12CR1
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions