missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LTBP2 Partial ORF (NP_000419, 1709 a.a. - 1818 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004053-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16157471

  • 349.00€ / 10µg
Estimated Shipment: 09-11-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene belongs to the family of latent transforming growth factor (TGF)-beta binding proteins (LTBP), which are extracellular matrix proteins with multi-domain structure. This protein is the largest member of the LTBP family possessing unique regions and with most similarity to the fibrillins. It has thus been suggested that it may have multiple functions: as a member of the TGF-beta latent complex, as a structural component of microfibrils, and a role in cell adhesion. [provided by RefSeq]

Sequence: LQPSELQPHYVASHPEPPAGFEGLQAEECGILNGCENGRCVRVREGYTCDCFEGFQLDAAHMACVDVNECDDLNGPAVLCVHGYCENTEGSYRCHCSPGYVAEAGPPHCT
Specifications

Specifications

NP_000419
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LQPSELQPHYVASHPEPPAGFEGLQAEECGILNGCENGRCVRVREGYTCDCFEGFQLDAAHMACVDVNECDDLNGPAVLCVHGYCENTEGSYRCHCSPGYVAEAGPPHCT
RUO
LTBP2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
4053
LTBP2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C14orf141/LTBP3/MSTP031
LTBP2
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions