missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Lubricin/PRG4 DuoSet ELISA
Click to view available options
Quantity:
1 Kit (for 5 Plates)
Unit Size:
1 set
Specifications
Specifications
| Assay Range | 15.60 to 1000 pg/mL |
| Product Type | Solid Phase Sandwich ELISA |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 10216 |
| Quantity | 1 Kit (for 5 Plates) |
| Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Synonym | bG174L6.2, bG174L6.2 (MSF: megakaryocyte stimulating factor), CACP, FLJ32635, HAPO, Hemangiopoietin, JCAP, lubricin, megakaryocyte stimulating factor, Megakaryocyte-stimulating factor, MSF, MSFproteoglycan 4, (megakaryocyte stimulating factor, articular superficial zoneprotein, camptodactyly, arthropathy, coxa vara, pericarditis syndrome), PRG4, Proteoglycan 4, Superficial zone proteoglycan, SZP, SZPHAPOarticular superficial zone protein |
| Target | Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction