missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LYSMD3 Full-length ORF (AAH58027.1, 1 a.a. - 127 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00116068-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16157283

  • 528.00€ / 25µg
Estimated Shipment: 09-11-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MAGRHQNRSFPLPGVQSSGQVHAFGNCSDSDILEEDAEVYELRSRGKEKVRRSTSRDRLDDIIVLTKDIQEGDTLNAIALQYCCTVYQNSSKKVQFLDRNTLSSKRKTDFTSFICSILFRTTGNFAS
Specifications

Specifications

AAH58027.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
40.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp686F0735/FLJ13542
LYSMD3
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
116068
LYSMD3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAGRHQNRSFPLPGVQSSGQVHAFGNCSDSDILEEDAEVYELRSRGKEKVRRSTSRDRLDDIIVLTKDIQEGDTLNAIALQYCCTVYQNSSKKVQFLDRNTLSSKRKTDFTSFICSILFRTTGNFAS
RUO
LYSMD3
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions