missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human/Mouse/Rat Phospho-JNK Pan Specific DuoSet IC ELISA

Product Code. 30021441
Change view
Click to view available options
Format:
2 96-well Plates
5 96-well Plates
Unit Size:
1 set
Product Code. Format unitSize
30021441 5 96-well Plates 1 set
30021087 2 96-well Plates 1 set
2 options
This item is not returnable. View return policy

Product Code. 30021441

Brand: R&D Systems™ DYC1387B5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human/Mouse/Rat Phospho-JNK Pan Specific DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human/Mouse/Rat Phospho-JNK Pan Specific DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human, mouse, rat JNK1/2/3.
TRUSTED_SUSTAINABILITY

Specifications

Product Type ELISA Kit
Quantity 1 Kit (for 5 Plates)
Species Human, Mouse, Rat
Conjugate Biotin
Assay Range 156.00 to 10000 pg/mL
Format 5 96-well Plates
For Use With (Application) ELISA
Synonym c-Jun N-terminal kinase 1, EC 2.7.11, EC 2.7.11.24, JNK1 alpha protein kinase, JNK1 beta protein kinase, JNK1JNK1A2, JNK-46, JUN N-terminal kinase, MAP kinase 8, MAPK 8, MAPK10, MAPK8, MAPK9, mitogen-activated protein kinase 8, mitogen-activated protein kinase 8 isoform JNK1 alpha1, mitogen-activated protein kinase 8 isoform JNK1 beta2, PRKM8, PRKM8JNK, protein kinase JNK1, SAPK1, SAPK1 alpha, SAPK1 beta, SAPK1JNK21B1/2, Stress-activated protein kinase 1, Stress-activated protein kinase JNK1
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.