missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human MPG - Ready-To-Use ELISA Kit (Colorimetric)

Product Code. 30238799 Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 30238799

Brand: Novus Biologicals™ NBP339179

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Colorimetric ELISA Kit

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.156 - 10 ng/mL (example only; lot dependent)
Assay Sensitivity 0.055 ng/mL (example only; lot dependent)
Detection Method Colorimetric
Immunoassay Kit Format Pre-coated 96T Strip Plate
Product Type ELISA Kit (Colorimetric)
Sample Type Tissue Homogenates, Other Biological Fluids
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 4350
Gene Symbol MPG
Quantity 1 Kit
Research Discipline Direct Reversal of DNA Damage
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym 3' end of the Mid1 gene, localized 68 kb upstream the humanzeta globin gene on16p, 3-alkyladenine DNA glycosylase, 3-methyladenine DNA glycosidase, AAG, ADPG, ANPG, APNG, CRA36.1, CRA36.1 (3-methyl-adenine DNA glycosylase), DNA-3-methyladenine glycosylase, EC 3.2.2.21, MDGalkyladenine DNA glycosylase, Mid1, N-methylpurine-DNA glycosylase, MPG, N-methylpurine-DNA glycosylasePIG16, PIG11, proliferation-inducing protein 11, proliferation-inducing protein 16
Target Human
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.