Learn More
Invitrogen™ Human MUC3 (aa 2768-2898) Control Fragment Recombinant Protein
Description
Highest antigen sequence indentity to the following orthologs: Mouse (30%), Rat (30%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-82409 (PA5-82409. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Specifications
Specifications
| Accession Number | Q02505 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 4584 |
| Name | Human MUC3 (aa 2768-2898) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | A630081J09Rik; intestinal mucin; Intestinal mucin-3 A; MUC3; MUC3A; MUC-3 A; mucin 3; mucin 3, intestinal; mucin 3 A; mucin 3 A, cell surface associated; Mucin-3; mucin-3 A; RIKEN cDNA A630081J09 gene |
| Common Name | MUC3 |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.