missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human nNOS (aa 1411-1425) Synthetic Peptide

Product Code. 11584840
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Product Code. Quantity unitSize
11584840 50 μg 50µg
1 options
This item is not returnable. View return policy

Product Code. 11584840

Brand: Invitrogen™ PEP190

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Synthetic Peptide

Human nNOS (residues 1411-1425) Synthetic Peptide, PEP-190, from Invitrogen.

NOS activity requires homodimerization as well as three cosubstrates (L-arginine, NADPH and O2) and five cofactors or prosthetic groups (FAD, FMN, calmodulin, tetrahydrobiopterin and heme). Several distinct NOS isoforms have been described and been shown to represent the products of three distinct genes. These include two constitutive Ca2+/CaM-dependent forms of NOS, including NOS1 (also designated ncNOS) whose activity was first identified in neurons, and NOS3 (also designated ecNOS), first identified in endothelial cells.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 4842
Species Human
Purification Method purified
Accession Number P29475
Common Name Human nNOS (aa 1411-1425)
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol NOS1
Lyophilized Lyophilized
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Sequence C N(1411) R L R S E S I A F I E E S K(1425)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.