missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ORC3L Full-length ORF (NP_862820.1, 1 a.a. - 712 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00023595-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16157512

  • 349.00€ / 10µg
Estimated Shipment: 09-11-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The origin recognition complex (ORC) is a highly conserved six subunits protein complex essential for the initiation of the DNA replication in eukaryotic cells. Studies in yeast demonstrated that ORC binds specifically to origins of replication and serves as a platform for the assembly of additional initiation factors such as Cdc6 and Mcm proteins. The protein encoded by this gene is a subunit of the ORC complex. Studies of a similar gene in Drosophila suggested a possible role of this protein in neuronal proliferation and olfactory memory. Alternatively spliced transcript variants encoding distinct isoforms have been reported for this gene. [provided by RefSeq]

Sequence: MATSSMSKGCFVFKPNSKKRKISLPIEDYFNKGKNEPEDSKLRFETYQLIWQQMKSENERLQEELNKNLFDNLIEFLQKSHSGFQKNSRDLGGQIKLREIPTAALVLGVNVTDHDLTFGSLTEALQNNVTPYVVSLQAKDCPDMKHFLQKLISQLMDCCVDIKSKEEESVHVTQRKTHYSMDSLSSWYMTVTQKTDPKMLSKKRTTSSQWQSPPVVVILKDMESFATKVLQDFIIISSQHLHEFPLILIFGIATSPIIIHRLLPHAVSSLLCIELFQSLSCKEHLTTVLDKLLLTTQFPFKINEKVLQVLTNIFLYHDFSVQNFIKGLQLSLLEHFYSQPLSVLCCNLPEAKRRINFLSNNQCENIRRLPSFRRYVEKQASEKQVALLTNERYLKEETQLLLENLHVYHMNYFLVLRCLHKFTSSLPKYPLGRQIRELYCTCLEKNIWDSEEYASVLQLLRMLAKDELMTILEKCFKVFKSYCENHLGSTAKRIEEFLAQFQSLDAETKEEEDASGSQPKGLQKTDLYHLQKSLLEMKELRRSKKQTKFEVLRENVVNFIDCLVREYLLPPETQPLHEVVYFSAAHALREHLNAAPRIALHTALNNPYYYLKNEALKSEEGCIPNIAPDICIAYKLHLECSRLINLVDWSEAFATVVTAAEKMDANSATSEEMNEIIHARFIRAVSELELLGFIKPTKQKTDHVARLTWGGC
Specifications

Specifications

NP_862820.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
108.7kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
LAT/LATHEO/ORC3
ORC3L
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
23595
ORC3L (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MATSSMSKGCFVFKPNSKKRKISLPIEDYFNKGKNEPEDSKLRFETYQLIWQQMKSENERLQEELNKNLFDNLIEFLQKSHSGFQKNSRDLGGQIKLREIPTAALVLGVNVTDHDLTFGSLTEALQNNVTPYVVSLQAKDCPDMKHFLQKLISQLMDCCVDIKSKEEESVHVTQRKTHYSMDSLSSWYMTVTQKTDPKMLSKKRTTSSQWQSPPVVVILKDMESFATKVLQDFIIISSQHLHEFPLILIFGIATSPIIIHRLLPHAVSSLLCIELFQSLSCKEHLTTVLDKLLLTTQFPFKINEKVLQVLTNIFLYHDFSVQNFIKGLQLSLLEHFYSQPLSVLCCNLPEAKRRINFLSNNQCENIRRLPSFRRYVEKQASEKQVALLTNERYLKEETQLLLENLHVYHMNYFLVLRCLHKFTSSLPKYPLGRQIRELYCTCLEKNIWDSEEYASVLQLLRMLAKDELMTILEKCFKVFKSYCENHLGSTAKRIEEFLAQFQSLDAETKEEEDASGSQPKGLQKTDLYHLQKSLLEMKELRRSKKQTKFEVLRENVVNFIDCLVREYLLPPETQPLHEVVYFSAAHALREHLNAAPRIALHTALNNPYYYLKNEALKSEEGCIPNIAPDICIAYKLHLECSRLINLVDWSEAFATVVTAAEKMDANSATSEEMNEIIHARFIRAVSELELLGFIKPTKQKTDHVARLTWGGC
RUO
ORC3L
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions