Learn More
Invitrogen™ Human p300 (aa 162-176) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP053
Additional Details : CAS : 69-74-9 Weight : 0.01000kg
Description
Human P300 (residues 162-176) Synthetic Peptide, PEP-053, from Invitrogen.
PEP-053 is a 14 amino acid synthetic peptide whose sequence corresponds to residues 139-151 of human p300. The sequence of this peptide is (amino to carboxy terminus): G139-T-S-G-P-N-Q-G-P-T-Q-S-T151-C (The C-terminal cysteine has been added to facilitate conjugation and is not a part of the native human p300 sequence)This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and p300, catalog # PA1-848. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Specifications
2033 | |
purified | |
Human p300 (aa 162-176) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
Q09472 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
EP300 | |
No preservative | |
50 μg | |
G139 - T - S - G - P - N - Q - G - P - T - Q - S - T151 - C |