missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human PAI-2 Recombinant Protein, PeproTech®

Product Code. 17890373
Click to view available options
Quantity:
10 μg
100 μg
2 μg
2 x 500 μg
250 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
2µg
500µg
Pack of 2
This item is not returnable. View return policy

Product Code. 17890373

Brand: Gibco™ 140062UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

140-06-1MG will be provided as 2 x 500 μg (140-06-500UG). Recombinant Human PAI-2 is a 46.5 kDa, nonglycosylated protein of 415 residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

PAI-2 is an inhibitory serpin expressed mainly in keratinocytes, activated monocytes, and placental trophoblasts. It exists predominantly as a 47 kDa, nonglycosylated, intracellular protein, which can be induced to be secreted as 60 kDa glycoprotein. The glycosylated and unglycosylated forms of PAI-2 are equally effective as inhibitors of urokinase-type plasminogen activator (uPA), the only established physiological target of this serpin. PAI-2 has a unique ability to form dormant polymers spontaneously and reversibly under physiological conditions. The physiological relevance of this property, which is neither a consequence of any mutation in the PAI-2 gene nor associated with any known disorder, is still unclear. However, it appears that the formation of intracellular, dormant polymers may be important for the controlled release of the inhibitor from PAI-2 producing cells. Plasma levels of PAI-2 are usually low or undetectable, except during pregnancy and in some forms of monocytic leukemia. Secretion of PAI-2 from the placenta normally occurs during the third trimester of pregnancy, and accounts for the dramatic increase in PAI-2 levels (up to 250 ng/ml), which are maintained at these levels until postpartum, and then rapidly decline. In addition to its vital role in protecting the placenta from degradation by uPA and/or uPA-activated proteases, PAI-2 has been shown to be essential for the prevention of metastatic spread of neck, lung and breast cancers. The beneficial effect of PAI-2 seen in these studies is presumed to stem from its ability to inhibit uPA-dependent cell dissemination. PAI-2 has also been reported to inhibit keratinocyte proliferation, and to participate in the innate immune response during viral infection.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P05120
For Use With (Application) Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 5055
Molecular Weight (g/mol) 46.5 kDa
Name Human PAI-2
Purification Method Purified
Quantity 2 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias HsT1201; Monocyte Arg-serpin; ovalbumin; PAI; Pai2; PAI-2; Pai2a; Placental plasminogen activator inhibitor; Planh2; Plasminogen activator inhibitor 2; plasminogen activator inhibitor 2 type A; Plasminogen activator inhibitor 2, macrophage; plasminogen activator inhibitor, type II; plasminogen activator inhibitor, type II (arginine-serpin); serine (or cysteine) peptidase inhibitor, clade B, member 2; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 2; serine (or cysteine) proteinase inhibitor, clade B, member 2; serpin B2; serpin family B member 2; serpin peptidase inhibitor, clade B (ovalbumin), member 2; SERPINB2; Urokinase inhibitor
Common Name SERPINB2
Gene Symbol SERPINB2
Biological Activity Determined by its inhibitory effect against single chain tPA induced cleavage of a chromogenic substrate in Imidazole Buffer at 37°C. Half maximal inhibition against 1.0 ug/ml of single chain tPA was obtained (at a concentration of 1.0 ug/ml)
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MEDLCVANTL FALNLFKHLA KASPTQNLFL SPWSISSTMA MVYMGSRGST EDQMAKVLQFNEVGANAVTP MTPENFTSCG FMQQIQKGSY PDAILQAQAA DKIHSSFRSL SSAINASTGNYLLESVNKLF GEKSASFREE YIRLCQKYYS SEPQAVDFLE CAEEARKKIN SWVKTQTKGKIPNLLPEGSV DGDTRMVLVN AVYFKGKWKT PFEKKLNGLY PFRVNSAQRT PVQMMYLREKLNIGYIEDLK AQILELPYAG DVSMFLLLPD EIADVSTGLE LLESEITYDK LNKWTSKDKMAEDEVEVYIP QFKLEEHYEL RSILRSMGME DAFNKGRANF SGMSERNDLF LSEVFHQAMVDVNEEGTEAA AGTGGVMTGR TGHGGPQFVA DHPFLFLIMH KITNCILFFG RFSSP
Content And Storage -20°C
Activity Determined by its inhibitory effect against single chain tPA induced cleavage of a chromogenic substrate in Imidazole Buffer at 37°C. Half maximal inhibition against 1.0 ug/ml of single chain tPA was obtained at a concentration of 1.0 ug/ml
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.