missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PAR1 ELISA Kit

Product Code. 17195129
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
This item is not returnable. View return policy

Product Code. 17195129

Brand: Invitrogen™ EH358RBX5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Human PAR1 quantitates human PAR1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human PAR1.

Thrombin is a serine protease that is involved in platelet aggregation and blood coagulation. It is cleaved from its precursor, prothrombin, and converts fibrinogen to fibrin in the final step of the clotting cascade. Thrombin mediates its regulatory effects by activating cell surface G-protein coupled receptors which include PAR-1, PAR-2 and PAR3.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AI482343; C HTR; Cf2r; coagulation factor II (thrombin) receptor; coagulation factor II receptor; coagulation factor II thrombin receptor; F2R; HTR; PAR1; PAR-1; protease activated receptor 1; protease-activated receptor 1; proteinase-activated receptor 1; Thrombin receptor; ThrR; TR; TRGPC
Assay Range 3 to 700 pg/mL
Assay Sensitivity 3 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 2149
Gene Symbol F2R
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target PAR1
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.