missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PD-ECGF ELISA Kit

Product Code. 17165139
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 10
Pack of 96
This item is not returnable. View return policy

Product Code. 17165139

Brand: Invitrogen™ EH364RBX5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Human PD-ECGF quantitates human PD-ECGF in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human PD-ECGF.

A platelet-derived endothelial growth factor (PD-ECGF), same as thymidine phosphorylase (TP) or gliostatin. In the presence of inorganic orthophosphate, it catalyses the reversible phospholytic cleavage of thymidine and deoxyuridine to their corresponding bases and 2-deoxyribose-1-phosphate. It is both chemotactic and mitogenic for endothelial cells and a non-heparin binding angiogenic factor present in platelets. It is also involved in transformation of fluoropyrimidines, cytotoxic agents used in the treatment of a variety of malignancies, into active cytotoxic metabolites.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 2900072D10Rik; ECGF; ECGF1; endothelial cell growth factor 1 (platelet-derived); Gliostatin; hPD-ECGF; MEDPS1; MNGIE; MTDPS1; OTTHUMP00000196767; OTTHUMP00000196770; PD TP; PDECGF; PD-ECGF; Pdgfec; platelet derived growth factor, endothelial cell; Platelet-derived endothelial cell growth factor; TdRPase; Thymidine phosphorylase; TP; TYMP
Assay Range 0.4 to 100 ng/mL
Assay Sensitivity 0.4 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 1890
Gene Symbol TYMP
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target Thymidine Phosphorylase
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.