missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Persephin Recombinant Protein, PeproTech®

Product Code. p-7246412
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
5µg
This item is not returnable. View return policy

Product Code. 17834673

Brand: Gibco™ 45012100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human Persephin is a disulfide-linked homodimer, composed of two 10.4 kDa polypeptide chains (194 total amino acid residues). Each chain contains seven conserved cysteine residues, one of which (Cys 64) is used for inter-chain disulfide bridging, and the others are involved in the intramolecular ring formation known as the cysteine knot configuration. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Persephin is a disulfide-linked, homodimeric, neurotrophic factor structurally related to GDNF, artemin, and neurturin. These proteins belong to the cysteine-knot family of growth factors that assume stable dimeric structures. Persephin signals through a multicomponent receptor system, composed of RET and one of four GFR alpha (alpha1-alpha4) receptors. The GFRalpha4 was first identified in chicken, and was later shown to be the preferential binding subunit for persephin. Persephin promotes the survival of ventral midbrain dopaminergic neurons and motor neurons after sciatic nerve oxotomy, and, like GDNF, promotes ureteric bud branching. However, in contrast to GDNF and neurturin, persephin does not support the survival of peripheral neurons.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O60542
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 5623
Molecular Weight (g/mol) 20.8 kDa
Name Human Persephin
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias Persephin; PSP; Pspn
Common Name PSPN
Gene Symbol PSPN
Biological Activity Human Persephin induces RET phosphorylation using a concentration range of 0.1-1.0 ng/ml Human Persephin binds to mammalian GFRa4 with the Kd of 100 pM. Other members of the GDNF family (Artemin, GDNF and Neurturin) do not bind to mammalian GFRa4.
Conjugate Unconjugated
Recombinant Recombinant
Sequence RALSGPCQLW SLTLSVAELG LGYASEEKVI FRYCAGSCPR GARTQHGLAL ARLQGQGRAH GGPCCRPTRY TDVAFLDDRH RWQRLPQLSA AACGCGG
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.