missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PHAP I (aa 79-110) Control Fragment Recombinant Protein

Product Code. 30207340
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30207340

Brand: Invitrogen™ RP103712

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

PHAP I is implicated in a number of cellular processes, including proliferation, differentiation, caspase-dependent and caspase-independent apoptosis, suppression of transformation (tumor suppressor), inhibition of protein phosphatase 2A, regulation of mRNA trafficking and stability in association with ELAVL1, and inhibition of acetyltransferases as part of the INHAT (inhibitor of histone acetyltransferases) complex. Plays a role in E4F1-mediated transcriptional repression.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P39687
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 8125
Name Human PHAP I (aa 79-110) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias Acid nuclear phosphoprotein 32 (leucine rich); acidic (leucine-rich) nuclear phosphoprotein 32 family, member A; acidic leucine-rich nuclear phosphoprotein 32 family member A; acidic nuclear phosphoprotein 32; acidic nuclear phosphoprotein 32 family member A; acidic nuclear phosphoprotein 32 family, member A; acidic nuclear phosphoprotein pp32; Anp32; ANP32A; C15orf1; cerebellar leucine rich acidic nuclear protein; hepatopoietin Cn; HPPCn; I1PP2A; inhibitor-1 of protein phosphatase-2 A; LANP; Leucine-rich acidic nuclear protein; leucine-rich acidic nuclear protein LANP; MAPM; Mapmodulin; MGC119787; MGC150373; PHAP1; PHAPI; Potent heat-stable protein phosphatase 2 A inhibitor I1PP2A; pp32; Putative HLA-DR-associated protein I; putative human HLA class II associated protein I; vacidic (leucine-rich) nuclear phosphoprotein 32 family, member A; W91701
Common Name PHAP I
Gene Symbol ANP32A
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GLEVLAEKCPNLTHLNLSGNKIKDLSTIEPLK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.