missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Phospho-p70 S6 Kinase (T389) DuoSet IC ELISA
Click to view available options
Quantity:
1 Kit (2 Plates)
1 Kit (5 Plates)
Unit Size:
1 set
Pack of 2
Pack of 5
Description
The Human Phospho-p70 S6 Kinase (T389) DuoSet IC ELISA quantifies phosphorylated human p70 S6 Kinase/S6K.
1 Kit (for 5 Plates)
Specifications
Specifications
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 6198 |
| Host Species | Human |
| Quantity | 1 Kit (5 Plates) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Target | p70 S6 Kinase/S6K |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction