missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Phospho-p70 S6 Kinase (T389) DuoSet IC ELISA

Product Code. 30021314
Click to view available options
Quantity:
1 Kit (2 Plates)
1 Kit (5 Plates)
Unit Size:
1 set
Pack of 2
Pack of 5
This item is not returnable. View return policy

Product Code. 30021314

Brand: R&D Systems™ DYC896B2

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Phospho-p70 S6 Kinase (T389) DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Phospho-p70 S6 Kinase (T389) DuoSet IC ELISA quantifies phosphorylated human p70 S6 Kinase/S6K.

1 Kit (for 5 Plates)

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) ELISA
Gene ID (Entrez) 6198
Host Species Human
Quantity 1 Kit (2 Plates)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Target p70 S6 Kinase/S6K
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.