missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Phospho-Tie-2 DuoSet IC ELISA

Product Code. 30021713
Click to view available options
Packaging Quantity:
15 Plates
2 Plates
5 Plates
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 30021713

Brand: R&D Systems™ DYC27202

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

contains the basic components required for the development of sandwich ELISAs to measure in cell lysates

The Human Phospho-Tie-2 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human Tie-2.
TRUSTED_SUSTAINABILITY

Specifications

Product Type ELISA Development Kit for Phosphorylated Intracellular Proteins
Quantity 1 Kit
Species Human
Conjugate Biotin
For Use With (Application) ELISA
Gene ID (Entrez) 7010
Packaging Quantity 2 Plates
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.