missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human PLA2R1 (NP_031392) VersaClone cDNA

Product Code. 18767748
Click to view available options
:
10 μg
This item is not returnable. View return policy

Product Code. 18767748

Brand: R&D Systems™ RDC2693

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.

  • Also known as: PLA2R; PLA2-R; PLA2IR; CLEC13C; PLA2G1R
  • PLA2R1 is a phospholipase A2 receptor.
  • It exists as both a transmembrane form and a soluble form.
  • The transmembrane receptor may play a role in clearance of phospholipase A2, thereby inhibiting its action.
  • Alternatively spliced transcripts encoding different proteins have been described.
  • The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
  • For reasearch use only. Not for use in humans.
TRUSTED_SUSTAINABILITY

Specifications

Gene hPLA2R1
Accession Number NP_031392
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Insert Type 4405 bp
Gene ID (Entrez) 22925
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.