missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Human Placenta Whole Tissue Lysate (Adult Whole Normal)
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
The Human Placenta Whole Tissue Lysate (Adult Whole Normal) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.
Specifications
Specifications
| For Use With (Application) | Western Blot |
| Tissue | Reproductive |
| Quantity | 1 mg |
| Storage Buffer | HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors |
| Test Specificity | Total Protein - Human Adult Normal Tissue: Placenta |
| Cellular Fraction | Whole |
| Tissue Condition | Normal |
| Cross Reactivity | Human |
| Host Species | Human |
| Concentration | 5 mg/mL |
| Show More |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction