missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant GDNF, ACF
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Unit Size:
1000µg
100µg
10µg
Description
- Glial cell line-derived neurotrophic factor (GDNF) is a neurotrophic factor and a member of the tumor growth factor (TGF)-beta superfamily
- The GDNF family of growth factors also includes neurturin, persephin, and artemin, which have seven conserved cysteine residues called cysteine-knot
- GDNF family ligands signal through binding to specific GDNF-family receptor-α (GFRα) co-receptors and activate the RET receptor tyrosine kinase
- Four different forms of GFRα co-receptors have been characterized (GFRα 1-4) out of which GDNF binds specifically to GFRα1 prior to forming a complex with RET
- GDNF is known to promote survival and morphological differentiation of midbrain dopaminergic neurons in both in vivo and in vitro studies and increase their high-affinity dopamine uptake
- GDNF has also been shown to have restorative effects on dying dopaminergic neurons in response to degenerative toxins
Specifications
Specifications
| Name | Astrocyte-derived trophic factor, ATF, ATF1, ATF2, Glial cell-derived neurotrophic factor |
| Quantity | 1000 μg |
| Cell Type | Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction