missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant IL-13
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Unit Size:
1000µg
100µg
10µg
Description
- Interleukin 13 (IL-13) is a cytokine important in type 2 immune responses and is expressed by T helper type 2 (Th2) cells and group 2 innate lymphoid cells (ILC2s)
- IL-13 binds a receptor composed of IL-4Ra and IL-13Ra1 or IL-13Ra2
- IL-13 receptor is expressed on B cells and promotes B cell proliferation, induces class switching to IgG4 and IgE, and functions in the recruitment and activation of IgE-producing B cells
- The receptor is also expressed on basophils, eosinophils, mast cells, endothelial cells, fibroblasts, monocytes, macrophages, respiratory epithelial cells, and smooth muscle cells
- Signaling through the IL-13 receptor activates the JAK/STAT and IRS-1/IRS-2 pathways
- In vivo, IL-13 has a role in resistance to extracellular helminth parasites by regulating gastrointestinal parasite expulsion, as well as in airway hyperresponsiveness, allergic inflammation, tissue remodeling, tumor cell growth, and fibrosis
Specifications
Specifications
| Name | Interleukin-13, NC30 |
| Quantity | 100 μg |
| Cell Type | B Cells; Hematopoietic Stem and Progenitor Cells; Monocytes |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction