missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Human Recombinant IL-13

Product Code. p-7217405
Encompass_Preferred
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Unit Size:
1000µg
100µg
10µg
This item is not returnable. View return policy

Product Code. 17139391

Brand: STEMCELL Technologies 78029

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Interleukin 13

  • Interleukin 13 (IL-13) is a cytokine important in type 2 immune responses and is expressed by T helper type 2 (Th2) cells and group 2 innate lymphoid cells (ILC2s)
  • IL-13 binds a receptor composed of IL-4Ra and IL-13Ra1 or IL-13Ra2
  • IL-13 receptor is expressed on B cells and promotes B cell proliferation, induces class switching to IgG4 and IgE, and functions in the recruitment and activation of IgE-producing B cells
  • The receptor is also expressed on basophils, eosinophils, mast cells, endothelial cells, fibroblasts, monocytes, macrophages, respiratory epithelial cells, and smooth muscle cells
  • Signaling through the IL-13 receptor activates the JAK/STAT and IRS-1/IRS-2 pathways
  • In vivo, IL-13 has a role in resistance to extracellular helminth parasites by regulating gastrointestinal parasite expulsion, as well as in airway hyperresponsiveness, allergic inflammation, tissue remodeling, tumor cell growth, and fibrosis

Specifications

Name Interleukin-13, NC30
Quantity 100 μg
Cell Type B Cells; Hematopoietic Stem and Progenitor Cells; Monocytes
Purity or Quality Grade ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.