missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Semaphorin 6A (NP_065847) VersaClone cDNA
Description
- SEMA6A semaphorin 6A [ Homo sapiens (human) ]
- Also known as: VIA; SEMA; HT018; SEMAQ; SEMA6A1
- The Semaphorin 6A insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- Sema6A is a transmembrane protein belonging to the semaphorin family.
- It is a cell surface receptor for PLXNA2.
- Sema6A promotes reorganization of the actin cytoskeleton and plays an important role in axon guidance in the developing central nervous system.
- It is required for normal granule cell migration in the developing cerebellum.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | hSEMA6A |
| Accession Number | NP_065847 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 3106 bp |
| Gene ID (Entrez) | 57556 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?