Learn More
Invitrogen™ Human SLC24A3 (aa 260-325) Control Fragment Recombinant Protein
Description
Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61020 (PA5-61020. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Specifications
Specifications
| Accession Number | Q9HC58 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 57419 |
| Name | Human SLC24A3 (aa 260-325) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | Na(+)/K(+)/Ca(2+)-exchange protein 3; NCK x 3; Slc24a3; sodium calcium exchanger; sodium/potassium/calcium exchanger 3; solute carrier family 24 (sodium/potassium/calcium exchanger), member 3; solute carrier family 24 member 3 |
| Common Name | SLC24A3 |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.