missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SMOC-2 ELISA Kit

Product Code. 17174733
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
This item is not returnable. View return policy

Product Code. 17174733

Brand: Invitrogen™ EH430RB

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Human SMOC-2 quantitates human SMOC-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human SMOC-2.

This gene encodes a member of the SPARC family (secreted protein acidic and rich in cysteine/osteonectin/BM-40), which are highly expressed during embryogenesis and wound healing. The gene product is a matricellular protein which promotes matrix assembly and can stimulate endothelial cell proliferation and migration, as well as angiogenic activity. Associated with pulmonary function, this secretory gene product contains a Kazal domain, two thymoglobulin type-1 domains, and two EF-hand calcium-binding domains. The encoded protein may serve as a target for controlling angiogenesis in tumor growth and myocardial ischemia. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number bA270C4A0.1; bA37D8.1; dJ421D16.1; DTDP1; MST117; MSTP117; MSTP140; Secreted modular calcium-binding protein 2; SMAP2; SMAP-2; SMOC2; SMOC-2; smooth muscle associated protein 2; smooth muscle-associated protein 2; SPARC related modular calcium binding 2; SPARC-related modular calcium-binding protein 2; thyroglobulin type-1 repeat containing protein
Assay Range 18 to 4,000 pg/mL
Assay Sensitivity 18 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 64094
Gene Symbol SMOC2
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target SMOC2
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.