missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human TACI/TNFRSF13B (NP_036584) VersaClone cDNA

Product Code. 18797159
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Product Code. Quantity unitSize
18797159 10 μg 10µg
1 options
This item is not returnable. View return policy

Product Code. 18797159

Brand: R&D Systems™ RDC1156

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Transmembrane Activator and CAML Interactor/Tumor Necrosis Factor Receptor Superfamily Member 13B. Human and mouse TACI share 54% amino acid sequence identity

The Human TACI/TNFRSF13B (NP_036584) VersaClone cDNA clone contains the complete ORF for TACI/TNFRSF13B/CVID, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The TACI/TNFRSF13B/CVID insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Order Info

Ships at ambient temperature

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_036584
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10mM Tris-Cl, pH 8.5
Insert Type 895bp
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg

For Reasearch Use Only; Not for Use in Humans

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.