missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human TNF-alpha Quantikine QuicKit ELISA

Product Code. 30001038
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Pack of 96
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001038 1 Kit Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001038 Supplier R&D Systems™ Supplier No. QK210

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

From the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

  • A one step, 80 minute solid phase ELISA designed to measure human TNF-alpha in cell culture supernates, serum, and plasma.
  • It contains E. coli-expressed recombinant human TNF-alpha and antibodies raised against the recombinant protein.
  • Results obtained for naturally occurring human TNF-alpha showed linear curves that were parallel to the standard curves obtained using the recombinant QuicKit™ standards.
  • These results indicate that this kit can be used to determine relative mass values for natural human TNF-alpha.
TRUSTED_SUSTAINABILITY

Specifications

Product Type ELISA
Specificity Natural and Recombinant Human TNF-alpha
Gene ID (Entrez) 7124
Host Species Human
Quantity 1 Kit
Storage Requirements Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Synonym APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Target TNF-alpha
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.