missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human TRANCE (RANKL) Recombinant Protein

Human TRANCE (RANKL) Recombinant Protein

Brand:  Gibco™ PHP0034

Additional Details : Weight : 0.02000kg

Product Code. 10727303

  • 445.00€ / Each
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Human TRANCE (RANKL) Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN

RANKL is a ligand for osteoprotegerin and functions as a key factor for osteoclast differentiation and activation. This protein was shown to be a dentritic cell survival factor and is involved in the regulation of T cell-dependent immune response. RANKL was shown to activate antiapoptotic kinase AKT/PKB through a signaling complex involving SRC kinase and tumor necrosis factor receptor-associated factor (TRAF) 6, which indicated this protein may have a role in the regulation of cell apoptosis.

Approved for shipment at Room Temperature or on Wet or Dry Ice.

Specifications

Specifications

Cell Culture
20 kDa
10 ug
Human
< 0.1 ng/μg
Approved for shipment on Wet or Dry Ice
Cytokines & Receptors
Lyophilized
Immunology, Bone Research, Signal Transduction, Apoptosis
98 %
8600
Sequential Chromatography
RANK Ligand
Store in freezer (-5 to -30°C).
10 μg
E. coli
Recombinant, Ligand
Other Proteins
Gibco™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.