missing translation for 'onlineSavingsMsg'
Learn More

Human TREM2 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

583.00€

Specifications

Antigen TREM2
Clone 1057114
Dilution Immunohistochemistry 5-25 ug/mL
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18781117
GREENER_CHOICE
View Documents
R&D Systems
MAB18282-SP
25 μg N/A
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.

Request a Price for this item here.

18743667
View Documents
R&D Systems
MAB18282-100
100 μg
583.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TREM2 Monoclonal antibody specifically detects TREM2 in Human samples. It is validated for Immunohistochemistry
Specifications

Specifications

TREM2
Immunohistochemistry 5-25 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
54209
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
1057114
Immunohistochemistry
Unconjugated
Mouse
Human
PLOSL2, TREM-2, Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2
NS0-derived recombiHumant human TREM-2 protein, His19-Ser174
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.