missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ULBP-3 ELISA Kit
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 10
Pack of 96
Description
Human ULBP-3 quantitates human ULBP-3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human ULBP-3.
UL16-binding protein 3 (ULBP3) is an extended member of the MHC class I superfamily. It does not contain the alpha 3 domain and lacks a transmembrane domain. ULBP3 binds and activates the KLRK1/NKG2D receptor and mediates natural killer cell cytotoxicity.
Specifications
Specifications
| Accession Number | ALCAN-gamma; N2DL3; N2DL-3; NKG2D ligand 3; NKG2D ligand3; NKG2DL3; RAET1N; Retinoic acid early transcript 1 N; UL16 binding protein 3; UL16-binding protein 3; ULBP; ULBP 3; ULBP3 |
| Assay Range | 1.3 to 300 pg/mL |
| Assay Sensitivity | 1.3 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 79465 |
| Gene Symbol | ULBP3 |
| Interassay CV | <12% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction