missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human UNQ3033 Full-length ORF (AAI00943.1, 1 a.a. - 174 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00284415-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16157429

  • 528.00€ / 25µg
Estimated Shipment: 10-11-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MTAEFLSLLCLGLCLGYEDEKKNEKPPKPSLHAWPSSVVEAESNVTLKCQAHSQNVTFVLRKVNDSGYKQEQSSAENEAEFPFTDLKPKDAGRYFCAYKTTASHEWSESSEHLQLVVTDKHDELEAPSMKTDTRTIFVAIFSCISILLLFLSVFIIYRCSQHSELRERKGREGE
Specifications

Specifications

AAI00943.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
46.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC119160/MGC119161/UNQ3033
VSTM1
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
284415
UNQ3033 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTAEFLSLLCLGLCLGYEDEKKNEKPPKPSLHAWPSSVVEAESNVTLKCQAHSQNVTFVLRKVNDSGYKQEQSSAENEAEFPFTDLKPKDAGRYFCAYKTTASHEWSESSEHLQLVVTDKHDELEAPSMKTDTRTIFVAIFSCISILLLFLSVFIIYRCSQHSELRERKGREGE
RUO
VSTM1
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions