missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human UQCRB Partial ORF (NP_006285, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007381-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16157325

  • 528.00€ / 25µg
Estimated Shipment: 09-11-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a protein which is part of the ubiquinol-cytochrome c oxidoreductase complex which contains ten nuclear-encoded and one mitochondrial-encoded subunits. The encoded protein binds ubiquinone and participates in the transfer of electrons when ubiquinone is bound. Mutations in this gene are associated with mitochondrial complex III deficiency. A pseudogene has been described on the X chromosome. [provided by RefSeq]

Sequence: MAGKQAVSASGKWLDGIRKWYYNAAGFNKLGLMRDDTIYEDEDVKEAIRRLPENLYNDRMFRIKRALDLNLKHQILPKEQWTKYEEENFYLEPYLKEVIR
Specifications

Specifications

NP_006285
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MAGKQAVSASGKWLDGIRKWYYNAAGFNKLGLMRDDTIYEDEDVKEAIRRLPENLYNDRMFRIKRALDLNLKHQILPKEQWTKYEEENFYLEPYLKEVIR
RUO
UQCRB
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
7381
UQCRB (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ92016/FLJ97033/QCR7/QP-C/QPC/UQBC/UQBP/UQPC
UQCRB
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions