missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human VISTA/B7-H5/PD-1H DuoSet ELISA

Product Code. 30021496
Click to view available options
Quantity:
1 Kit (for 5 Plates)
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 30021496

Brand: R&D Systems™ DY712605

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For determination of natural and recombinant human V-domain Ig Suppressor of T cell activation.

The Human VISTA/B7-H5/PD-1H DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human VISTA/B7-H5/PD-1H.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Solid Phase Sandwich ELISA
Quantity 1 Kit (for 5 Plates)
Species Human
Binding Target VISTA/B7-H5/PD-1H
Assay Range 23.40 to 1500 pg/mL
Format 96-well
Gene ID (Entrez) 64115 (Human); 74048 (Mouse); 690899 (Rat); 102116468 (Cynomolgus Monkey)
Sample Type Cell Culture Supernatants, Serum, EDTA Plasma, Heparin Plasma
Synonym 4632428N05Rik, B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, Dies1, Gi24, PD1H, PD-1 H, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR
Kit Contents 96 well Micro Plates, PBS, Plate Sealers, Reagent Diluents Concentrate 2, Stop Solution, Substrate Solution, Wash Buffer
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Show More Show Less

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.