missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZMYND19 Full-length ORF (NP_612471.1, 1 a.a. - 227 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00116225-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16157323

  • 349.00€ / 10µg
Estimated Shipment: 09-11-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ZMYND19 is a MYND zinc finger domain-containing protein that binds to the C terminus of melanin-concentrating hormone receptor-1 (MCHR1; MIM 601751) (Bachner et al., 2002 [PubMed 12208518]), and to the N termini of alpha-tubulin (TUBA1; MIM 191110), and beta-tubulin (TUBB; MIM 191130) (Francke et al., 2005 [PubMed 16039987]).[supplied by OMIM]

Sequence: MTDFKLGIVRLGRVAGKTKYTLIDEQDIPLVESYSFEARMEVDADGNGAKIFAYAFDKNRGRGSGRLLHELLWERHRGGVAPGFQVVHLNAVTVDNRLDNLQLVPWGWRPKAEETSSKQREQSLYWLAIQQLPTDPIEEQFPVLNVTRYYNANGDVVEEEENSCTYYECHYPPCTVIEKQLREFNICGRCQVARYCGSQCQQKDWPAHKKHCRERKRPFQHELEPER
Specifications

Specifications

NP_612471.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
52.8kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MIZIP/RP11-48C7.4
ZMYND19
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
116225
ZMYND19 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTDFKLGIVRLGRVAGKTKYTLIDEQDIPLVESYSFEARMEVDADGNGAKIFAYAFDKNRGRGSGRLLHELLWERHRGGVAPGFQVVHLNAVTVDNRLDNLQLVPWGWRPKAEETSSKQREQSLYWLAIQQLPTDPIEEQFPVLNVTRYYNANGDVVEEEENSCTYYECHYPPCTVIEKQLREFNICGRCQVARYCGSQCQQKDWPAHKKHCRERKRPFQHELEPER
RUO
ZMYND19
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions