missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human HUNK, GST Tag Recombinant Protein
This HUNK Recombinant Human Protein (full length) was expressed in insect cells.
524.00€ - 7580.00€
Specifications
Accession Number | NP_055401 |
---|---|
For Use With (Application) | Kinase Assay |
Molecular Weight (g/mol) | 105.9. kDa |
KinaseFamily | CAMKL Kinase Family |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17168745
|
Invitrogen™
A32892 |
100 μg |
2435.00€
100µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15920303
|
Invitrogen™
A30974 |
10 μg |
524.00€
10µg |
Estimated Shipment: 30-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17118795
|
Invitrogen™
A33377 |
1 mg |
7580.00€
1mg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
HUNK serine/threonine kinase or hormonally up-regulated Neu-associated kinase is a member of the mitogen-activated protein kinase family which is essential for mammary tumor metastasis. HUNK deletion in mice did not alter viability, fertility, longevity, or propensity to develop tumors and it is also dispensable for murine development.- Validated for use with the LanthaScreen™ Eu Kinase Binding assay
- Available in multiple pack sizes for convenient use
Specifications
NP_055401 | |
105.9. kDa | |
Full Length | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol. | |
Baculovirus | |
Full Length, Recombinant, Active |
Kinase Assay | |
CAMKL Kinase Family | |
Human | |
GST Tag | |
Dry Ice | |
Kinases & Inhibitors | |
Liquid |