Learn More
Hamilton™ HVC Standard Ceramic Valves
Designed to provide superior performance with salt solutions.
314.00€ - 503.00€
Specifications
Description Ceramic HV valve
View More Specs
Products 5
Product Code Brand For Use With (Equipment) Product Type Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Product Type Price Quantity & Availability  
11543314
View Documents Promotion Details
Hamilton™
5208-01
PSD/2 and 3 Syringe Drive Modules 3-2 Model, 3 Ports, Aluminum Housing
314.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 18-04-2024
to see stock.
11593394
View Documents Promotion Details
Hamilton™
6763-01
PSD/2 and 3 Syringe Drive Modules 3-2 Model, 3 Ports, Stainless Steel Housing
503.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 18-04-2024
to see stock.
11583404
View Documents Promotion Details
Hamilton™
6764-01
PSD/2 and 3 Syringe Drive Modules 3-5 Model, 3 Ports, Stainless Steel Housing
440.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 18-04-2024
to see stock.
11573344
View Documents Promotion Details
Hamilton™
7991-01
PSD/4 and 8 Syringe Drive Modules 3-5 Model, 3 Ports, Aluminum Housing
344.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 18-04-2024
to see stock.
11513484
View Documents Promotion Details
Hamilton™
8063-01
PSD/4 and 8 Syringe Drive Modules T 3-2 Model, 3 Ports, Aluminum Housing
486.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 18-04-2024
to see stock.
Description
Description
  • Rigid ceramic fluid path is resistant to scratching from salt crystals
  • Provides superior life and performance
  • Exceptionally easy to service and modify
  • Maximizes flexibility and value
  • Ideal for low pressure applications
  • All HVC valves have a thread depth of 1/4 in.-28 UNF
Specifications
Specifications
Ceramic HV valve