missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Characterized Fetal Bovine Serum, Australia Origin
Enhances the growth of cells
231.00€
Specifications
Country of Origin | Australia |
---|---|
Form | Frozen |
Product Type | Fetal Bovine Serum |
Sterility | Sterile |
Sterilization Method | Sequential Triple-filtered, 100 nm |
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
12614207
|
Cytiva
SH30084.02IR |
Gamma Irradiated |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Description
- Considered to be free of BSE and FMD
- USDA safety-tested
- Tested for endotoxin and hemoglobin levels
- Sterile filtered using true pool technology
- Characterized FBS is filtered through sequential, 100 nm pore-size rated filters
- Gamma irradiation further reduces the risk of adventitious viruses present in serum
Specifications
Australia | |
Fetal Bovine Serum | |
Sequential Triple-filtered, 100 nm | |
≤ 25 mg/dL | |
USDA Safety Tested |
Frozen | |
Sterile | |
≤ 25 EU/mL | |
HyClone™ |