Learn More
Cytiva HyClone™ HyCell-STEM™ Media
Product Code. 15276057
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15276057 100 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Hyclone Cell Culture products now a part of GE Healthcare.
Product Code. 15276057 Supplier Cytiva Supplier No. SR30003.01
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Hyclone Cell Culture products now a part of GE Healthcare.
Includes
HyCell-STEM kit includes 6x supplement and DMEM/F12 to create a complete media. Basic fibroblast growth factor must be added to the completed media at time of use.
Requires
bFGF (10ng/ml) must be added to the media at time of use.
Complete media for culturing human embryonic stem cells (hESCs) and induced pluripotent stem (hiPS) cells.
HyCell-STEM Media is designed to help generate large numbers of human embryonic and induced pluripotent stem cells with fewer passages and is intended for use with stem cells on feeder layer cultures. HyCell-STEM is provided as a 6x supplement that is added to DMEM/F12 to create a complete media. Basic fibroblast growth factor is added to the completed media at time of use.
Highlights:
  • Increase rate of expansion
  • Sustain healthy stem cell morphology
  • Maintain stemness
  • Preserve pluripotency
  • Improved post-thaw recovery
HyCell-STEM Media is intended for use with stem cells on feeder layer cultures.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Stem Cell Supplement Media
For Use With (Application) Stem Cell Culture
Quantity 100 mL