Learn More
HYDRANAL™ - Coulomat AF 7, For coulometer AF7, Honeywell Fluka™
Product Code. 15674490 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 L
6 x 1 L
Unit Size:
1 L
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15674490 1 L 1 L
15684490 6 x 1 L Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Product Code. 15674490 Supplier Honeywell-Hydranal Supplier No. 348291L
Request Bulk or Custom Format
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
for coulometer AF7
Advantages of Hydranal Coulometric Reagents
  • Easy to use
  • High accuracy for trace amounts of water
  • Stable conditions of the titration vessel
  • Broad product range
  • Long shelf life (up to five years)
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material HYDRANAL™-Coulomat AF 7
CAS 288-32-4, 7446-09-5, 67-56-1
Quantity 1 L
Flash Point 11°C (51.8°F)
Packaging Glass bottle
UN Number UN1230