missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine hydrate solution, 78-82% (iodometric), Honeywell Fluka™

Product Code. p-7121616 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
This item is not returnable. View return policy

Product Code. 15681650

Brand: Honeywell-Fluka 18412250ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 10217-52-4
Molecular Formula H4N2
Molecular Weight (g/mol) 32.05
MDL Number MFCD00149931
InChI Key OAKJQQAXSVQMHS-UHFFFAOYSA-N
Synonym hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739
PubChem CID 24654
ChEBI CHEBI:35511
IUPAC Name hydrazine
SMILES NN

Specifications

Assay Percent Range 78 to 82%
Linear Formula NH2NH2 · H2O
UN Number UN2030
Beilstein 878137
Formula Weight 50.06
Percent Purity 78 to 82% (iodometric)
Grade Organics
Packaging Plastic Bottle
Quantity 250 mL
Assay acidimetric
Chemical Name or Material Hydrazine hydrate solution, 78-82% acidimetric
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.